Use your mouse or the controls below to move the slide
MOVE
ZOOM
HINT
Count all of the cells in the squares outlined in red. Use ZOOM to increase the magnification.
Enter your cell count for 4 squares
cells × 2 (dilution factor) × 10,000 cells/mL
dead cells × 2 (dilution factor) × 10,000 cells/mL
Ready to give up manual cell counting?
See how to convince your lab to get an automated cell counter >
Improve your cell counting accuracy
Download the white paper >
Tips & Tricks for Cell Culture
Download the Gibco™ Cell Culture Basics handbook >
See how to convince your lab to get an automated cell counter >
Improve your cell counting accuracy
Download the white paper >
Tips & Tricks for Cell Culture
Download the Gibco™ Cell Culture Basics handbook >