"How many events do you need to acquire?"
Assay variation can be a major source of error, therefore it is critical to understand the statistics behind rare event analysis. The cytometry friendly Gaussian distribution cannot be applied to the analysis of such a low number of events (the term "rare" is given to events with a frequency of 0.01% or less) and the best approach is to use Poisson statistics (Fig.1). Good experimental practice suggests to keep Coefficient of Variation (CV) below 5%, and thus the number of events to acquire should be defined in order to maintain the lowest CV possible and in any case below 5%.
The number of events that satisfy a given criterion (e.g they are positive for a marker, P) is defined as
$P=R/N$
where
$N$=total events
$R$=events that meet a given criterion
and
$0<=P<=1$
As with all statistical distribution,
the variance (V) is defined as
$V(R) = NP(1-P)$
The standard deviation (SD) is
$SD=√V=√(NP(1-P))$
and the coefficient of variation is
$CV=1/√V$
Figure 1. Poisson statistics defines the probability that a number of events will occur in a fixed interval of time/space/volume.
Use the tool below to help define the ideal number of events to acquire for the expected frequency of rare population.
Step 1: Input the expected frequency in the cell below; i.e. to identify a cell population that represents 0.01% of the total, input frequency is 0.001 (one positive event every ten thousands)
The number of events that satisfy a given criterion (e.g they are positive for a marker, P) is defined as
$P=R/N$
where
$N$=total events
$R$=events that meet a given criterion
and
$0<=P<=1$
As with all statistical distribution,
the variance (V) is defined as
$V(R) = NP(1-P)$
The standard deviation (SD) is
$SD=√V=√(NP(1-P))$
and the coefficient of variation is
$CV=1/√V$
Figure 1. Poisson statistics defines the probability that a number of events will occur in a fixed interval of time/space/volume.
Step 2: Use the Events (N) cells below to input the total number of events, CV% cells will automatically refresh allowing the identification of the ideal total events number or the interval required to achieve a CV below 5%; i.e. to identify a cell population that represents 0.01%, five million events should be acquired.
Events (N)
Positive (R)000000
Proportion (P)000000
Variance (V)000000
SD000000
CV%000000
Where:
Number of events$$N$$
Positive events$$R$$
Proportion (frequency)$$P=R/N$$
Variance$$V=NP(1-P)$$
Standard Deviation$$SD=√V$$
Coefficient of Variation %$$CV=(SDx100)/R$$
Learn how Attune Flow Cytometers can help you count cells and identify populations.
Attune Flow Cytometers