Invitrogen E-Gel Power Snap Plus Electrophoresis System – all-in-one imaging and analysis for high-throughput workflows.
The integrated design offers:
Fast and safe workflow using Invitrogen E-Gel technology, Invitrogen SYBR Safe DNA Gel Stain, and blue-light transilluminator.
Convenience of gel-running and documentation on the same small device.
Easy analysis on both the device and on the Thermo Fisher Connect Platform with Invitrogen iBright image analysis software.
The Invitrogen SuperScript IV UniPrime One-Step RT-PCR System is the latest addition to the Invitrogen SuperScript product family. It combines Invitrogen SuperScript IV Reverse Transcriptase (RT) with the novel Invitrogen UniPrime RT-PCR Master Mix for ease of use and unmatched one-step RT-PCR performance.
  • Simplified workflow—universal primer annealing at 60°C decreases optimisation time.
  • Easy pipetting—colored buffers for visual tracking of reaction setup and direct sample loading on gels.
  • Enabled automation—two-phase hot-start activation mechanism for room-temperature setup and benchtop stability for up to 24 hours.
  • Superior performance—high yields and reliable target detection even with challenging RNA samples.
The Applied Biosystems VeritiPro Dx Thermal Cycler is an in vitro diagnostic (IVD) endpoint thermal cycler designed to amplify nucleic acids from human-derived specimens using polymerase chain reaction (PCR). It is classified as a US FDA Class I Medical Device. It conforms to (EU) 2017/746 requirements and is CE‑IVD labeled in Europe, making it the only IVDR-compliant thermal cycler. The VeritiPro Dx Thermal Cycler delivers precision temperature controls with ramp rate speeds of 6.0°C/sec. The premium performance enables reliable and fast PCR-based workflow results.
For In-Vitro Diagnostic Use.
Invitrogen SuperScript IV RT-LAMP Master Mix is a reverse transcription loop-mediated isothermal amplification (RT-LAMP)–based solution for faster and simpler detection of viral RNA. It detects DNA targets, enabling a wider range of applications for research and surveillance of pathogens (Incl. SARS-CoV-2).
New customers save 50%learn more
The TaqMan Cells-to-CT Express Kit allows for rapid gene expression analysis directly from cultured cells without the need for RNA purification. With a simplified workflow and high sensitivity, this kit provides RT-qPCR results in approximately 70 minutes.
Features of the TaqMan Cells-to-CT Express Kit include:
  • Extraordinary ease and speed—samples ready for RT‑qPCR in as little as 5 minutes; convenient one‑tube RT master mix featuring SuperScript IV VILO for two‑step RT‑qPCR
  • No tedious RNA purification—no columns, heating, or centrifugation
  • Verified workflows—complete set of reagents optimized to work efficiently right out of the box
  • Environmentally friendly—no hazardous chemicals and fully REACH compliant reagents; fewer components and pipetting steps, reducing plastic cost and waste
  • Right sized kits for automation—available in 2500 reaction size that includes both lysis and RT‑qPCR component
SuperScript IV Template Switching RT Master Mix is designed for high template switching efficiency in cDNA synthesis reactions.
Features of SuperScript IV Template‑Switching RT Master Mix include:
  • Formulated for high quality full‑length cDNA synthesis
  • Includes proprietary enhancer to maximize template switching efficiency
  • Exceptional sensitivity for easy detection of low abundance targets (as low as single cell or 2 pg total RNA)
  • Cell lysis buffer included for direct cDNA synthesis without RNA purification
The Thermo Scientific EquiPhi29 DNA Amplification Kit includes all required components for fast, sensitive, and efficient isothermal DNA amplification, which includes rolling circle amplification (RCA) and multiple displacement amplification—whole genome amplification (MDA‑WGA).
Features include:
  • Fast DNA amplification in less than two hours
  • Sensitivity from 1 fg of input DNA
  • Maximal yield of 17 µg amplified DNA
  • Compatibility with different types of sample input material (e.g., purified DNA, liquid media culture, agar plate colonies)
  • Compatibility with various downstream applications such as DNA sequencing, digestion with restriction enzymes, cell‑free DNA enrichment, and cell‑free protein expression
  • All required components for DNA amplification in RCA or MDA‑WGA reactions
Thermo Scientific PCR plastics, Sustain Series products, which bring together precision, reliability, and sustainability
The biobased polypropylene resin used in the Thermo Scientific PCR plastics, Sustain series products is manufactured from a second‑generation, bio‑circular feedstock (waste and residual oils) under a mass balance approach.
These ISCC PLUS‑certified, biobased PCR plastics go beyond providing accurate and reproducible PCR data, they represent our commitment to integrating more sustainable practices into our manufacturing processes, without compromising on product quality or performance.
Together, let’s contribute to a greener future, one PCR experiment at a time.
Invitrogen Lyo-ready Bst DNA Polymerase is a proprietary mutant of Bst DNA polymerase, large fragment, with a fast reaction speed and increased sensitivity and tolerance to inhibitors. Lyo-ready Bst DNA polymerase provides maximum flexibility to optimize your LAMP reaction and work with various types of pathogens, including human adenovirus, measles, SARS-CoV-2, and other pathogens.
Invitrogen One Shot LB Agar Plates
  • Ready-to-use bacterial growth media
  • Pre-poured LB Agar Plates (LB Miller’s formulation) with glass beads
  • Convenient for immediate use, with a longer shelf life for flexibility and reduced waste
Enzymes for In vitro Transcription
Enzymes for In vitro Transcription cover the entire RNA synthesis workflow from template preparation to post‑transcriptional modifications.
Invitrogen GeneArt Services Dashboard
For over 20 years, Invitrogen GeneArt Gene Synthesis clones and GeneArt Strings DNA Fragments have been critical tools for research and discovery. Now, we are introducing the easiest way to access these services and more.
The Invitrogen GeneArt Services Dashboard includes:
  • Intuitive design
  • Industry‑leading gene optimisation
  • Order tracking
  • Quick pricing estimator
  • Detailed quote generation
qPCR and PCR plastics selection tool
Select your real-time PCR (qPCR) or thermal cycler (PCR) instrument and easily find the plates, tube strips, single tubes, caps, seals, adhesive films, and accessories compatible with your instrument.
PCR Enzymes Selection Tool
Quickly find PCR Enzymes that are best suited for your research applications. Spend more time on your experiments than looking for an enzyme that you need.
Gene Synthesis Vector Selection Tool
Take advantage of our selection guide to review our most popular Invitrogen vectors for gene synthesis. The tool allows you to quickly identify a vector that best suits your needs.
Thermo Scientific FastDigest Enzyme Selection Tool
Through the FastDigest collection, you have access to one of the largest collections of restriction enzymes (176) in the industry and the first simplified restriction enzyme system using a universal buffer.
Competent cells decision tree
Use the component cell selection guide to help determine which competent cells are best suited for your cloning experiments.
Invitrogen GeneArt Seamless Cloning and Selection Tool
This is a tool for assembling multiple DNA Fragments to build large and seamless clones.
GeneArt Seamless Cloning and GeneArt Gibson Assembly Kits allow for the simultaneous assembly of up to 15 DNA fragments to create precise, very large constructs with no unwanted sequences in highly efficient reactions, saving 3-4 weeks versus traditional Restriction Enzymes cloning methods.
E-Gel Precast Gel and E-Gel Ladder Selection Tool
Find the right E-Gel precast gel and the E-Gel ladder for your nucleic acid separation and analysis needs with this tool.
Whether you are assessing your PCR product, purifying a DNA fragment of interest for cloning, or preparing DNA fragments for your Next Generation Sequencing (NGS), simply select the desired parameters and see the E-Gel and ladder recommendations instantly.
Reverse Transcriptase Selection Guide
Use this selection guide to search for the right reverse transcriptase, based on application, format, reaction time, and other experimental parameters to meet your cDNA synthesis needs.
School of molecular biology
Whether you want to review the basics, gain more in-depth knowledge, or discover our latest research tools out educational hub provides reliable technical content designed for new and experienced molecular biologists alike
Molecular biology resource library
This page contains valuable links and supporting resources for standard molecular biology techniques, as well as specialty applications. It also provides easy access to a large variety of Thermo Scientific webinars, videos, articles, and more.
Molecular cloning education
Learn about the foundations of molecular cloning and recombinant DNA technology. Find resources suitable for researchers at multiple levels, covering restriction enzymes, traditional cloning, and competent cells as fundamental research tools in various applications.
Synthetic biology learning center
Synthetic biology comprises the next generation of genetic engineering tools. It combines molecular biology and systems biology with engineering principles to design biological systems and bio-factories. The aim is to create improved biological functions that meet current and future challenges.
Molecular biology spotlight articles
Get meaningful results at the bench with practical tips and tricks!
This collection features various molecular biology applications for PCR, cloning, reverse transcription, and more.
We highlight the latest scientific breakthroughs and constantly look for new advancements to help fulfilling all your research needs.