Complete this form and a technical specialist will contact you
Looking for a more affordable source of Protein A resin? Thermo Scientific MabCaptureC affinity matrix is a high‑performance Protein A chromatography resin, specifically designed to help improve the productivity and efficiency of your antibody purification process. Featuring high capacity and a highly cross‑linked agarose backbone, the MabCaptureC resin is your Protein A resin of choice for efficient process‑scale purification of monoclonal antibodies.
Learn more ›
Advantages of the MabCaptureC Affinity Matrix
- High dynamic binding capacity (DBC): >50 g/L at 4.8‑minute residence time
- Excellent alkaline stability (0.2 M NaOH >100 cycles) allowing for efficient cleaning
- Scalable and ready for use in commercial manufacturing
How to request a sample
- Submit your details: Complete the form with your basic information.
- Consult with a specialist: Our consultants will contact you to understand your needs.
- Enjoy your sample: Evaluate your resin and contact us if you need additional support.
Additional Resources
Terms and conditions:
* Participation is limited to researchers and purchasing personnel working in the life science industry in the US (excluding Puerto Rico), Canada, South America, Europe, the Middle East, and Africa who are 21 years of age or older, who request and evaluate a sample for a new molecule or workflow by December 31, 2025, and who subsequently place a qualified order requesting the discount with a Thermo Fisher Scientific technical sales specialist within 6 months of sample shipment. Sample sizes are available in 25 mL and 50 mL Thermo Scientific POROS GoPure Column, Thermo Scientific CaptureSelect RoboColumn
columns, Thermo Scientific POROS RoboColumn columns, Thermo Scientific MabCaptureC RoboColumn columns, and Thermo Scientific POROS GoPure 96-well screening plate formats. Limit one sample of each type per customer. Customer must include a unique identifier connected to samples and agree to evaluate the sample before placing an order to receive the discounted price on their next order. Customers can use the discount only once per type. Discount applies only to affinity resins up to 5 L and/or polish resins up to 25 L. Other items in an order are not eligible for discount. Discount applies to the list price in effect at the time the order is received by Thermo Fisher Scientific. Offer terms, including end date of promotion, may be changed by Thermo Fisher Scientific without prior or further notice. Cannot be combined with other discounts or promotions. Health care professionals and government officials may not participate in this promotion. By requesting a sample, you warrant that you are not prohibited by employment, contract, or law from accepting a sample. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
POROS: Pharmaceutical Grade Reagent. For Manufacturing and Laboratory Use Only.
MabCaptureC: For Research Use or Further Manufacturing. Not for diagnostic use or direct administration in humans or animals.
*MiniChrom and RoboColumn are registered trademarks of Repligen.