Webinar #3: Enabling neurological disease research via DNA fragment analysis on the new SeqStudio Genetic Analyzer
This is the third topic in the Applied Biosystems SeqStudio Webinar Series, ‘Sanger sequencing and fragment analysis made easy’.
Repetitive DNA sequences comprise half of the human genome and include at least 1.6 million short tandem repeats (STRs) with unknown functions and disease implications. Recent genome-wide studies have illuminated thousands of associations of such repeat elements with changes in gene expression. In addition, over 30 Mendelian disorders are known to be caused by triplet repeat expansions. Thus, there is a rapidly growing appreciation that repetitive “dark” DNA is discoverable, discernible and diagnostic.
In this webinar, we will discuss:
- How reliable, long-read AmplideX® PCR technology can be combined with the Applied Biosystems SeqStudio Genetic Analyzer to make routine the detection of established and emerging repeat DNA markers in an easy-to-use assay system
- Data showing sensitive and accurate analyses of GC- and AT-rich repeat expansions and length polymorphisms associated with fragile X syndrome, ALS/FTD, myotonic dystrophy, Huntington’s disease and Alzheimer’s disease
- The implications of high-resolution repeat DNA genotyping for scientific and clinical research of neurodevelopmental, neurodegenerative and neuromuscular disorders
Other topics in the webinar series include:
- Introduction to SeqStudio applications (#1)
- Detect low-level somatic mutations in FFPE samples using Sanger sequencing (#2)
- Genome editing workflow facilitated by the Thermo Fisher Scientific portfolio solution (#4)
- Cell line authentication using STR analysis (#5)
![Gary Latham, PhD](images/gary_latham.jpg)
About the presenter
Gary Latham, PhD
Senior Vice President, Research and Development
Asuragen
Dr. Latham is Senior Vice President of R&D for Asuragen, a molecular diagnostics company focused on genetics and oncology. Dr. Latham joined Asuragen when it was founded in 2006, and he currently oversees the company's research, assay development, partnered R&D, and bioinformatic teams. He guided the development of AmplideX® PCR technology for fragile X syndrome and other DNA repeat disorders, and DNA- and RNA-based QuantideX® NGS products for oncology applications. Dr. Latham is an inventor on 12 issued US patents, has received >$13M in research grants from the NIH and the Cancer Prevention and Research Institute of Texas (CPRIT), and has authored >40 peer-reviewed publications.
Complete the form to register for the webinar.
For Research Use Only. Not for use in diagnostic procedures.