Discover the kit's performance dataset—download the CISA-INIA validation report
The Applied Biosystems VetMAX African Swine Fever Virus Detection Kit is a single-well duplex real-time PCR for the detection of African Swine Fever Virus (ASFV) DNA in serum, whole blood, and tissue samples of pigs and wild boars using TaqMan primer and probes. It enables detection in individual samples or pools of up to 10 samples.
The kit is sensitive and specific for the detection of ASFV DNA, allowing for reliable and rapid screening during outbreaks and monitoring situations. It has been validated by the European Union Reference Laboratory and has been used since 2015 during outbreaks in Eastern Europe for clinical sign confirmation and detection of domestic and wild pigs affected by ASFV.
Complete the form below and receive the validation report performed by the Centro de Investigacion en Sanidad Animal (CISA-INIA), the European Union Reference Laboratory for ASF (EURL-ASF).