POROS CaptureSelect AAV bioprocess columns are pre-packed chromatography columns specifically designed for the purification of a broad range of AAV serotypes. The use of these columns eliminates the need for column packing, column qualification, and sanitization, thereby saving time, resources, and costs.
These pre-packed process scale columns are suitable for cGMP manufacturing and have the following features:
- Prepared and packed in a classified ISO 7 environment
- Qualified in terms of performance and pressure flow characteristics
- Suitable for direct connection to standard chromatography systems
- Regulatory support documentation for both the column hardware and the resin
- Designed to be used for multiple cycles to improve process cost efficiency
Please fill out the request form to receive your quote.