Choose from our TaqMan SARS-CoV-2 2.0 assays
Find the right assay for your needs from our evolved menu.
TaqMan SARS-CoV-2 Fast PCR Combo Kit 2.0
- Compatible with raw saliva (no preservatives)
- Utilizes RNase P as an internal control
- Fast, direct-to-PCR workflow from raw saliva to results in ~2 hours
TaqMan SARS-CoV-2 RNase P Assay 2.0
- Recommended for use with RNA extracted from nasopharyngeal and anterior nasal swabs
- Utilizes RNase P as an internal control
- Go from extracted RNA to results in ~3 hours
Please fill out the form, and a sales representative will be in touch.
For research use only. Not for use in diagnostic procedures.