A high volume electroporation system designed for cell therapy development and manufacturing
The Gibco CTS Xenon Electroporation System delivers consistently high transfection performance in a volume up to 25 mL at a time with exceptional cell viability and recovery. It can be used to deliver DNA, RNA and protein payloads like Gibco CTS HiFi Cas9 Protein, designed to improve specificity while maintaining high editing efficiency. The intuitive programmable interface, process flexibility, sterile single-use consumables, and available 21 CFR Part 11 compliance compatible software upgrade allow the system to seamlessly scale with your cell therapy process from development through clinical manufacturing.
To request a quote for the CTS Xenon Electroporation System, please provide your information below.
(*Indicates a mandatory field)
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products.