Discover cutting edge technology for the purification of DNA, RNA, proteins, and cells
Building on decades of experience and customer feedback, the KingFisher Apex delivers unrivaled flexibility and performance.
New features include:
  • Modern icon-based touchscreen for ease of use
  • Expanded heating and cooling controls to maintain sample integrity
  • Library of validated protocols for multiple applications
  • Elute in 10 µL of buffer for more concentrated sample output
  • Two UV lights to safeguard against contamination
  • Compatible with a wide range of Applied Biosystems MagMAX sample prep kits
(*Indicates a mandatory field)