Thermo Scientific Armadillo 96-well PCR Strip Plates combine the rigidity of a polycarbonate frame with thin-walled polypropylene wells to provide superior thermal cycling performance under all conditions without warping. The specially designed warp-resistant frame makes Armadillo PCR Strip Plates the ultimate choice for high-throughput PCR with automated sealing and lyophilization. Armadillo PCR Strip Plates can be easily and safely broken into 8-tube strips by hand to provide maximum scalability while minimizing waste.
If you are developing PCR-based assays, the versatility of the Armadillo PCR Strip Plates makes them the perfect complement to our oligos and lyo-ready enzymes.
Request your free sample pack today by completing the form below.