Enable scalability and performance with the Thermo Scientific DynaDrive Single‑Use Bioreactor. Please fill out the form, and we'll contact you shortly.
Designed to meet the evolving needs of the biopharmaceutical industry, the Thermo Scientific DynaDrive Single‑Use Bioreactor (S.U.B.) offers exceptional scalability, performance, and reliability. Enabling scale‑up from research and development to full‑scale production, the DynaDrive S.U.B. is designed to provide the flexibility and efficiency required to help streamline your processes and help you achieve your goals
Key benefits of the DynaDrive S.U.B.
- A wide range of working volumes: Available from 1 L to 5,000 L
- Consistent performance: Uniform mixing and oxygen transfer rates across all scales helps maximize cell density to support fed-batch and perfusion processes
- Ease of scale-up: Consistent design principles across different sizes (aspect ratio, modified elephant ear impeller, product contact materials)
- Greater performance and scalability: The DynaDrive S.U.B. can decrease the cost of production by up to 25% through a reduction in equipment, materials, consumables, and labor going far beyond the limits of legacy single‑use bioreactor systems
- Network reliability: A leading single‑use manufacturing network for reliable BPC sourcing
Are you ready to elevate your bioprocessing?
Fill out this form to request more information.