Cell protection
Thermo Scientific centrifuges, CO2 incubators and biological safety cabinets are technologically advanced to help keep your samples safe from contamination, protect your next discovery and help advance your research
Centrifuges
- Biocontainment with ClickSeal Biocontainment Lids, validated by Public Health England
- Auto-Lock Rotor Exchange Technology gives full chamber access for better cleaning, minimizing risk of contamination
- Sorvall BIOS model: stainless steel exterior option and Auto-Door & Auto-Lid to reduce hands-on operation
Watch video fullscreen:
ClickSeal Biocontainment Lids
CO2 incubators
- Proven 180°C dry heat sterilization cycle
- ISO Class 5 HEPA (High Efficiency Particulate Air) filters in chamber
- Optional 100% pure copper interiors provide natural protection
- Separated water reservoir—covered integrated humidity reservoir maximizes relative humidity without condensation ensuring a dry inner chamber
- Cell Locker System for enhanced protection in an isolated environment
Watch video fullscreen:
Cell Locker System
Biological safety cabinets
- SmartFlow Plus—independent intelligent DC motors for downflow and inflow monitoring and control in real time to maintain balanced airflow
- DAVe Plus—Unique digital airflow verification system validates protection by monitoring airflow velocities with independent pressure sensors
- Cross-Beam UV Plus—Self-adjusting UV gemicidal light for better contamination control
- Smart-Clean Plus window—fully opening front hinged window design for simplified cleaning
Watch video fullscreen:
SmartFlow Plus technology
Contact us for help in selecting the right centrifuge, CO2 incubator or biological safety cabinet for your cell culture lab
Advanced cell culture technology
Thermo Scientific centrifuges, CO2 incubators and biological safety cabinets offer cutting-edge technology with touch screen user interfaces and connectivity options to advance your next discovery.
Centrifuges
Technologies for productivity, ergonomics and sustainability
- Secure, push-button rotor exchange in as little as 3 seconds with Auto-Lock technology
- General Purpose Pro Centrifuges: intuitive touchscreen interface for simple and fast programming
- Improved ergonomics with long-lasting Fiberlite Carbon Fiber Rotors having 15 years warranty
- Automatic door opening/closure functions
- Compact footprint and reduced noise
- USB port for easy data transfer
Read SmartNote about our Auto-Lock technology ›
CO2 incubators
Technologies for optimal growth conditions
- THRIVE active airflow technology—total recovery of all critical growth parameters in 10 minutes or less following a 30 second door opening
- Probes and gas sensors positioned in the chamber respond quickly to any deviations in environmental conditions
- Tri-Gas Incubators—Variable Oxygen Control incubators are uniquely designed to optimise growth of mammalian cells in an environment that closely mimics in vivo conditions
Watch video fullscreen:
THRIVE technology
Biological safety cabinets
Technologies for simplified operation, ergonomics and documentation requirements
- Intuitive user interface (touchscreen as standard for Herasafe 2030i model)
- Electronic front sash—With push button simplicity, the front window automatically positions itself
- Adjustable ergonomic armrests and optional removable gel-filled comfort pads for enhanced user experience
- Enhanced information management—can be connected to remotely monitor and preserve important performance data (Herasafe 2030i series)
View SmartNote on the user interface ›
Contact us for help in selecting the right centrifuge, CO2 incubator or biological safety cabinet for your cell culture lab
Reliability for your peace of mind
Thermo Scientific centrifuges, CO2 incubators and biological safety cabinets are reliable and built to last, in order for you to concentrate on your next discovery and help your lab work to flourish.
Centrifuges
- Over 60 years of experience in centrifugation
- Quality products, manufactured at an ISO 13485 certified production site in Osterode am Harz, Germany
- Durable Fiberlite carbon fiber rotors with a 15-year warranty
- Global service and support
CO2 incubators
- Over 50 years of experience with the development and production of CO2 incubators
- Quality products, manufactured in Langenselbold, Germany and Marietta, Ohio, USA
- To date, we have built more than 100,000 Heracell models
- Global service and support
Biological safety cabinets
- Over 50 years of experience with clean air products
- Quality products, manufactured in Langenselbold Germany
- Global service and support
Learn more ›
Culture with product variety
Thermo Scientific centrifuges, CO2 incubators and biological safety cabinets, offer a variety of equipment options to meet individual application needs helping to drive your next discovery.
Centrifuges
Application:- Small to large scale
- Unmatched rotor portfolio for application flexibility
- Unique rotor designs enabling simultaneous microplate, conical tube and bottles spinning
Capacity:
- Microcentrifuges
- Small benchtop centrifuges
- General purpose centrifuges
- Large capacity centrifuges
- Our centrifugation portfolio offers sample processing possibilities from 0.2 mL and up to 16 L
Options/Configurations/Accessories:
- Centrifuge tubes
- Rotors
- ClickSeal Biocontainment lids
CO2 incubators
Application:- Small to large scale
- Normal to enhanced protection
- Oxygen control (tri-gas) models available
Capacity:
Wide variety of sizes and stackable options (note: different model configurations available):
- Midi 40 CO2 incubator: 40L
- Heracell 150i/240i: 150/240L
- Heracell VIOS or Forma Steri-Cycle 160i/250i: 165/255L
- Forma Steri-Cult: 322L
- Large-Capacity Reach-In Incubators: 821L
Options/Configurations/Accessories:
- Variety of model configurations
- Customization options available
- Cell culture consumables
- Cell culture media
Biological safety cabinets
Application:- Class II models, Cytotoxic safety cabinets (for applications requiring the highest personnel, product and environmental protection) or clean benches
Capacity:
- Different sizes available: 3-foot, 4-foot, 5-foot, 6-foot
- Lowered work surface option
Options/Configurations/Accessories:
- Height adjustable floor stands, trays, hose connections, optional particle monitoring, cross-beam UV Plus
- Ergonomic stool
- Ergonomic gel-filled armrest
Contact us for help in selecting the right centrifuge CO2 incubator or biological safety cabinet for your cell culture lab.