• 1
    SELECT
    Expression System
    Show all
    Posttranslational modifications
    Application
    Speed to protein
    (expression duration)
    Protein yield
    2
    FIND Products
    Organism
    System
    Cells
    Media
    Transfection / Delivery
    Expression vector
    Protein labeling
    Additional protein expression products
    Additional workflow products
  • ×
    Human
    Human (HEK293)
    Full
    Low or no-yield proteins,
    Structural biology,
    Reagent production,
    Intracellular proteins,
    Viral proteins,
    Membrane proteins,
    Fc fusion proteins,
    Antibody production
    2-7 days
    Up to 5 g/L
    Human
    Expi293 PRO
    Expi293 PRO Cells
    Expi293 PRO Expression Medium
    Expi293 PRO Transfection Kit
    pcDNA 3.4,
    pcDNA 3.3,
    pTT5
    Expi293 Met (-) Protein Labeling Kit,
    L-Methionine (Methyl-13C), L-Selenomethionine
    Expi293 Expression System,
    Expi293F Cells,
    Expi293 Expression Medium,
    ExpiFectamine 293 Transfection Kit,
    Expi293 GnTI- Expression System Kit,
    Expi293F GnTI- Cells,
    Expi293 Inducible Expression System Kit,
    Expi293F Inducible Cells,
    Expi293 Inducible GnTI- Expression System Kit,
    Expi293F Inducible GnTI- Cells,
    Expi293 Met (-) Expression Medium
    Antibody Expressing Control,
    Nalgene Erlenmeyer Flasks
  • ×
    Hamster
    Hamster (CHO)
    Full
    Antibody production,
    Bispecifics proteins,
    Fc fusion proteins
    7-14 days
    Up to 3 g/L
    Hamster
    ExpiCHO
    ExpiCHO-S,
    ExpiCHO-S Cells (cGMP Banked)
    ExpiCHO Expression Medium
    ExpiFectamine CHO Transfection Kit
    pcDNA 3.4,
    pcDNA 3.3,
    pTT5
    –
    Freedom ExpiCHO-S Kit
    Antibody Expressing Control,
    Nalgene Erlenmeyer Flasks
  • ×
    Insect
    Insect (Sf9, Sf21)
    Partial
    Intracellular proteins,
    Toxic proteins
    3-5 days
    Up to 300 mg/L
    Fall armyworm (Spodoptera frugiperda)
    ExpiSf
    ExpiSf9
    ExpiSf CD Medium
    ExpiFectamine Sf Transfection Reagent
    pFastBac 1, pFastBac Dual, pFastBac HT
    Sf-900 II SFM, no met,
    no cys
    Bac-to-Bac TOPO N-His,
    Bac-to-Bac TOPO C-His,
    Bac-to-Bac TOPO HBM
    Nalgene Erlenmeyer Flasks
  • ×
    Yeast
    Yeast
    Partial
    Low-complexity proteins
    3-5 days
    Up to 10 g/L
    Pichia pastoris
    PichiaPink,
    EasySelect Pichia
    PichiaPink Strain Set, GS115, X-33
    PichiaPink Media Kit, YPD
    Neon Transfection System
    pPINK (LC and HC), pPINKα-HC,
    pPICZ A, B & C, pPICZα
    –
    Multi-Copy Pichia,
    Pichia EasyComp
    Nalgene Erlenmeyer Flasks
  • ×
    Bacterial
    Bacterial
    None
    Basic expression
    1-2 days
    Up to 10 mg/L
    E. coli
    Champion pET, T7,
    pBAD
    BL21, BL21-AI
    MagicMedia,
    Luria Broth (LB)
    None required
    Champion pET, pBAD, pRSET
    –
    pTrc A, B & C
    Nalgene Erlenmeyer Flasks