b'Cloud-enabled Cloud-enabled Automation-readySimpliAmpThermal CyclerMiniAmpThermal Cycler Automated Thermal Cycler Do you need an intuitive interface? Do you want an instrument with just the featuresPCRDo you want to place your Do you train new technicians often? needed for routine PCR? instrument on a robotic platform Do you want to access your instrument remotely? Do you want to access your instrument remotely? now or in the future? Elegantly simple and preciseRoutine PCR, elevatedDesigned for easy robotic integration 96 reactions96 reactions384 reactions 4.0C/sec3.5C/sec3.5C/sec3-zone VeriFlex Block on 96-well system 3-zone VeriFlex Block Noneon MiniAmp Plus modelYes Yes YesFind out more at thermofisher.com/thermalcyclers 23'