b'All CytoScan arrays are hybrid-SNP chromosomal microarrays (CMAs) that contain large numbers of bothSNP probes and non-polymorphic probes. Hybrid-SNP arrays, with more than 99% genotype accuracy,boost confidence in breakpoint determination and enable independent confirmation of copy number eventsthroughout the entire genome. Applied Biosystems microarrays for research applications in cytogenetics analysisPrenatal Postnatal Liquid tumors Solid tumorsFresh/Frozen FFPECytoScan OptimaCytoScan 750KCytoScan HDOncoScanCytoScan HT-CMACytoScan XONFigure 2. Applied BioSystems microarrays enable a wide range of cytogenetics research application areas.Hybrid-SNP arrays can detect these types of geneticDownload whitepapers:conditions1 for research applications: Learn how high-resolution Suspected uniparental disomy (UPD), whole genome UPD,SNP arrays detect inherited cnLOH, or AOH chromosomal anomaliesMosaicismZygosity Cytogenetic applicationsConsanguinity With advances in human genetic analysis technologies, Parent of origin (requires parental genomic analysis) various types of causative genetic aberrations associated with disorders can be detected thus helping provide valuable Twin-twin or maternal cell contamination insights into chromosomal abnormalities in constitutional andAllele-specific changes oncological samples.Sample heterogeneity, clonal diversity Prenatal genetic testing helps clinical researchers study Genomic contamination congenital anomalies or genetic disorders in the fetus.Postnatal genetic testing helps clinical researchers study SNP analyses are also used for: congenital anomalies, diseases, and developmental delays in Breakpoint determinationcases following birth.Mendelian consistency checkingProduct of Conception (POC) genetic testing of fetal tissue after the loss of a pregnancy helps clinical researchers study Confirmation of CNV events if the miscarriage was caused by a chromosomal anomaly.Oncological genetic testing of tumor samples helps clinical researchers study chromosomal aberrations to guide investigations of potential cancer treatments.5'