b'Imaging and spectrophotometryThe Thermo Scientific VitroEase Buffer Screening Kit providesFeatures of the VitroEase Buffer Screening Kit:preformulated buffers and detergents for a comprehensive screening strategy to identify optimal protein conditions for single particle Ease of usepreformulated and ready to be spiked into protein cryo-electron microscopy (cryo-EM) sample analysis. samples with minimal sample dilution Convenientcolor-coded caps that correlate with a color-coded Cryo-EM sample screening with various buffers and additives is donescreening strategyto evaluate protein behavior with the aim of identifying the optimal conditions in which a protein is stable, randomly oriented, and equally Comprehensiveoptimized screening strategy encompasses two distributed in ice.rounds of screening with 24 grids and troubleshooting suggestions Flexiblewide range of common buffers and detergents sufficient for sample optimization of 400 grids Validatedcomponents and screening strategy verified with single particle cryo-EM analysisOrdering informationProduct Cat. No.VitroEase Buffer Screening Kit A49856Learn more at thermofisher.com/em-sample-prepContents Inside the Cell//Issue No. 11 15'