b'Protein research Continue the western workflow virtual tour: Mini Gel Tank and Mini Blot ModuleFamiliarize yourself with the Invitrogen Mini Gel Tank, InvitrogenKey highlights:Mini Blot Module, and Invitrogen PowerEase Touch Power Supplies through our 3D virtual tour. Side-by-side tank configurationallows simultaneous visualization of two gelsDiscover the many unique benefits of our mini-gel chamber system and Forward-facing well configurationfor easy sample loadingsee how easy it is to use for electrophoresis and western blot transfers. View the PowerEase Touch Power Supply software interface to see the Less running and transfer buffer requiredtwo separate pre-programmed and customizable method options available. chambers; use only the buffer needed for each gel or transfer Compatiblefits all Invitrogen precast mini-gels and Invitrogen SureCast hand-cast mini-gels Half-inch transfer buffer chamberrequires only 220 mL of methanol-based transfer buffer per blot, reducing buffer and disposal costs Robust electrodes, sturdy steel platesfor highly efficient and reliable western transfersTake the tour at thermofisher.com/minigeltank22 Inside the Cell//Issue No. 11Contents '