b'Multiplex analysisThe Invitrogen Luminex xMAP INTELLIFLEX platform is the most recent addition to the xMAP line of instruments. The advantages of this new platform include a compact footprint with an integrated PC and touchscreen; next-generation optics with user-friendly instrument updates; and data output that is compatible with analysis applications on the Thermo Fisher Connect Platform.The INTELLIFLEX platform is available in two configurationsthe Invitrogen Luminex xMAP INTELLIFLEX System and the Invitrogen Luminex xMAP INTELLIFLEX DR-SE System. The DR-SE version has a second reporter channel and a microplate side eject feature for seamless integration with liquid handling workstations.Features of INTELLIFLEX instrumentsLuminex xMAP Luminex xMAPINTELLIFLEX DR-SE System INTELLIFLEX SystemReporter laser(s)532 nm (green)532 nm (green) 405 nm (violet)Side ejector portPlate format (96-well, 384-well)Small footprint including PC (58.4 x 61 x 76.2 cm)21 CFR compliance (2021)Ordering informationProduct Cat. No.Luminex xMAP INTELLIFLEX System APX2020Luminex xMAP INTELLIFLEX DR-SE System APX2021Discover more at thermofisher.com/luminexinstrumentsContents Inside the Cell//Issue No. 11 9'