b'Cell analysis and imagingEnhanced imaging experience with EVOS Optimized Light CubesInvitrogen EVOS and Invitrogen Countess second-generation LED light cubes (2.0) take cell imaging to the next level for even better publication-quality images. These interchangeable light cubes deliver precise control and plug-and-play capability.Features of the optimized light cubes include: Improved opto-mechanical design More uniform illumination across field of view (FOV) Excellent multi-FOV stitching performance Spectrally optimized optical components Improved spectral fidelity across channels to eliminate undesired bleed-through Maximized signal-to-background ratio even with dim samples Compatible with legacy EVOS Light Cubes and Invitrogen Countess FL Automated Cell CountersOrdering informationProduct Dye Cat. No.EVOS Light Cube, TagBFP 2.0 TagBFP AMEP4968 EVOS Light Cube, Cy5.5 2.0 Cy5.5 AMEP4973Learn more at thermofisher.com/evoslightcubesContents Inside the Cell//Issue No. 12 11'