b'Western workflowThe Invitrogen PowerEase Touch 600W Power Supply joins the PowerEase Touch 120W and 350W Power Supplies, adding another level of power and enabling users to conduct isoelectric focusing experiments (Table 1).Like its siblings, the PowerEase Touch 600W Power Supply has a bright, LCD touchscreen interface, which provides easy entry of up to 100 custom programs, or selection of preprogrammed protocols for Invitrogen protein gels and gel transfers. Table 1. Comparison of PowerEase Touch Power Supply models.120 W (basic) 350 W600 W (universal)(high current)Mini gel runs 4 12 12Mini gel transfers 2 8 8Midi gel runs 2 8 8Midi gel transfers No 4 4Isoelectric focusing (IEF) gels No No YesPower output 1120 W 1350 W 1600 WCurrent output 1500 mA 1 mA3 A 1 mA3 AVoltage output 2300 V 2300 V 2500 VExplore your options at thermofisher.com/powereaseContents Inside the Cell//Issue No. 12 5'