b'Cell analysis and imagingCelleste 6 Image Analysis Softwarenow available for EVOS M5000 and M7000 Imaging SystemsInvitrogen Celleste 6 Image Analysis Software is a full-featured image analysis suite designed for biological applications that enables users of the Invitrogen EVOS M5000 and M7000 Imaging Systems to seamlessly capture, process, segment, classify, measure, analyze, and share images and data.The multi-channel analysis (MCA) tool featured in version 6 utilizes preconfigured algorithms and analysis templates, which have been trained on representative data, to optimally segment and classify images from a range of common cell-based assays. Simply choose the app that corresponds to your assay of interest and follow the wizard-based workflow steps from image to data generation. Bioapplications include cell viability, apoptosis, cell cycle, live/dead cell analysis, transfection efficiency, immunohistochemistry analysis, migration tracking, wound healing, adipogenesis, and cytoskeletal disruption.Ordering informationProduct Cat. No.Celleste 6 Image Analysis Software AMEP4942To learn more, visit thermofisher.com/celleste12 Inside the Cell//Issue No. 12Contents '