b'ImmunoassaysWe now have a complete, comprehensive solution for researchers who want to investigate the 10 hallmarks of cancer using Invitrogen ProcartaPlex multiplex assays. With 46 additional targets and 6 extra panels, researchers can choose from any of these important hallmarks, including exosomal characterization, to perform cancer research.Hallmarks of cancerHuman Immune Checkpoint(EPX370-15846-901, 37-plex) Human Growth Factor Panel (EPX110-12170-901, 11-plex)Human Immuno-Oncology BDNF, EGF, FGF-2, HGF, LIF, NGF beta,Checkpoint Panel 1PDGF-BB, PIGF-1, SCF, VEGF-A, VEGF-D (EPX14A-15803-901, 14-plex) CD27, CD28, CD137 (4-1BB), GITR, HVEM, BTLA, CD80, CD152 (CTLA4), IDO, LAG-3, Human Exosome Characterization PanelPD-1, PD-L1, PD-L2, TIM-3(EPX060-15845-901, 6-plex) CD81, CD63, CD9,VLA-4, Syntentin-1,Human Immuno-Oncology Cytochrome C Evading Tumor- Checkpoint Panel 2growth promoting (EPX140-15815-901, 14-plex)suppressors inammationMICA, MICB, Perforin, ULBP-1, ULBP-3, ULBP-4, Arginase-1, CD73 (NT5E), CD96 (Tactile), E-Cadherin, Nectin-2, PVR, Human Cell Death PanelDeregulating Evading Siglec-7, Siglec-9(EPX040-15843-901, 4-plex)immuneHSP70, HSP90, Dkk-1, HMGB-1 (HMG-1) cellular destruction Human Immuno-Oncology energetics Checkpoint Panel 3Human Apoptotic Cell Clearance Panel(EPX090-15820-901, 9-plex)(EPX120-15816-901, 12-plex)B7-H6, CD276 (B7-H3), CD47 (IAP), AXL, CD36, CALR (CRT), Gas6, LOX-1,Sustained CD48 (BLAST-1), CD134 (OX40), ICOS MBL, Mer (MERTK), CD31 (PECAM-1),proliferative Invasion and Ligand (B7-H2), TIMD-4, S100A8/A9, PAI-1 (Serpin), uPAR, RAGE, TYRO3 signaling metastasis VISTA (B7-H5)GenomeHuman Cell Proliferation and Metastasis Human Angiogenesis Panel 1instability and Replicative Panel 1(EPX180-15806-901, 18-plex)mutation immortality (EPX130-15841-901, 13-plex) Angiopoietin-1, BMP-9, EMMPRIN,AFP, ADAM12, Beta-catenin, CD171 Follistatin, HB-EGF, LYVE-1, TIE-2,Resisting Inducing (NCAM-L1), CD66f (PSBG-1), Galectin-1, Syndecan-1, EGF, FGF2, G-CSF (CSF-3) ,cell death angiogenesis hCG, HO-1, IGFBP-1, N-Cadherin, HGF, IL-8 (CXCL8), Leptin, PDGF-BB,Nectin-4, PAPP-A, Thrombomodulin 1CD31 (PECAM-1), VEGF-A, VEGF-DHuman Cell Proliferation and Metastasis Human Angiogenesis Panel 2Panel 2(EPX030-15807-901, 3-plex)(EPX120-15842-901, 12-plex)Angiogenin, Angiostatin, Endostatin MIP-4 (CCL18), Haptoglobin, IGFBP-3, Human Cell Proliferation PanelHGFR (c-Met), MIA, VE-Cadherin, (EPX080-15844-901, 8-plex)Cathepsin D, CEA (CEACAM-5), Periostin IGF-2, Midkine (MK), SPARC (Osteonectin), TrkB,(OSF-2), Beta-2-microglobulin (B2M), HSP27, Glypican-1 (GPC1), EpCAM, HSP60 IGFBP-2, EGFR (ErbB1)Ordering informationProduct Cat. No.Cell Death 4-Plex Human ProcartaPlex Panel EPX040-15843-901Cell Proliferation 8-Plex Human ProcartaPlex Panel EPX080-15844-901Cell Proliferation and Metastasis 13-Plex Human ProcartaPlex Panel 1 EPX130-15841-901Cell Proliferation and Metastasis 12-Plex Human ProcartaPlex Panel 2 EPX120-15842-901Exosome Characterization 6-Plex Human ProcartaPlex Panel EPX060-15845-901Immuno-Oncology Checkpoint 9-Plex Human ProcartaPlex Panel 3 EPX090-15820-901Choose from the above preconfigured panels or design your own Mix & Match Oncology panel using the ProcartaPlex Panel Configuator at thermofisher.com/findluminex.Find out more at thermofisher.com/procartaplexContents Inside the Cell//Issue No. 12 19'