b'Cell analysis and imagingInvitrogen Countess 3 and Countess 3 FL Automated Cell Counters have updated software features that include the ability to support 21 CFR Part 11 compliance. This FDA requirement addresses the transition from paper record keeping to electronic record keeping. The Countess 3 and 3 FL Automated Cell Counters use security, audit, and electronic signature (SAE) software to support electronic record keeping compliance. The purchase of a license key (Cat. No. A51025) unlocks the SAE mode on the Countess 3 instrument and allows customers to connect the instrument to their SAE server. Additionally, updated features in the latest software update include refined FCS file reporting (enabling greater usability), optimized lighting settings, and expanded profile functions, including saving preferred dilution settings. These features are all freely accessible via software download and instrument update.Ordering informationProduct Cat. No.Countess 3 21 CFR Part 11 Software Module A51025Find out more at thermofisher.com/countesscfrContents Inside the Cell//Issue No. 12 9'