b'Western workflowProtein gels welcome packs for protein expression analysis Two money-saving protein gels welcome packs have been designed for use with any protein expression application, such as transient protein production using the Gibco Expi293 Expression System, enabling you to analyze your expression results quickly and easily. These welcome packs provide all necessary gels, buffers, and reagents as well as a fast 12-minute gel stain to visualize electrophoresis results. An Invitrogen gel tank is included at essentially no extra cost.The gels provided in the Invitrogen Bolt Mini Bis-Tris Welcome Pack and the Invitrogen NuPAGE Midi Bis-Tris Welcome Pack can be run in as little as 2035 minutes and stained in only 12 minutes.Key benefits: Bolt Mini Bis-Tris Welcome Pack for Protein Expression (412%, 12-well) Protein resolutionthe 412% polyacrylamide gradients of the Bolt Mini Bis-Tris Plus gels (12-well) and the higher-throughput NuPAGE Midi Bis-Tris gels (20-well) deliver sharp, well-resolved bands Rapid time-to-resultsBolt Mini Bis-Tris Plus gels can be run in 20 minutes, and NuPAGE Midi Bis-Tris gels can be run in 35 minutes; quick, 12-minute staining protocol using Invitrogen SimplyBlue SafeStain Conveniencethe Invitrogen Mini Gel Tank offers easy, side-by-side sample loading into the larger-volume, wedge-shaped wells of the Bolt Mini Bis-Tris Plus gels; run up to 40 samples in the Invitrogen SureLock Tandem Midi Gel Tank, or purchase 26-well gels for even higher throughput Two-in-one gel tank included at no extra costperform wet tank protein transfers for western blot analysis in the same tank at roomNuPAGE Midi Bis-Tris Welcome Pack for Protein Expression (412%, 20-well)temperature with the purchase of separately available blot modulesLearn more at thermofisher.com/proteingelwelcome6 Inside the Cell//Issue No. 12Contents '