b'Nucleic acid quantificationPrecise and accurate nucleic acid quantification is a critical step in many DNA and RNA research workflows. Success of downstream analyses (e.g., sequencing, PCR, transfection) often depend on the use of an appropriate amount of sample.The Invitrogen Qubit Flex Fluorometer is a benchtop instrument designed for accurate and sensitive quantification of DNA, RNA, and protein samples. This one-of-a-kind instrument offers researchers the ability to measure up to 8 samples simultaneously. The updated instrument firmware (version 1.5.0) provides researchers with greater flexibility and functionality.Enhanced firmware features: Flexibility to calibrate fewer than 8 wellsmaking the workflow faster for low-throughput users Fluorometer mode with blue or red excitation for users withcustom assaysAbility to export data as a PDF for customers requiring a non-editable results document Ordering informationProduct Cat. No.Qubit Flex Fluorometer Q33327Qubit Flex NGS Starter Kit Q45893Qubit Flex Quantitation Starter Kit Q45894Download the updated firmware at thermofisher.com/qubitfirmwareContents Inside the Cell//Issue No. 12 21'