b'E-Gel precast gels Using precast agarose gels can simplify the nucleic acid electrophoresis workflow. There are no gels to pour, buffers to make, staining or destaining steps to perform, or gel boxes to assemble. Just load your samples and run.Invitrogen E-Gel CloneWell II gels have a double comb design that enables recovery of purified DNA for downstream applications. You can obtain higher yields of purified DNA from the recovery wells in an E-Gel CloneWell II gel than you would by extracting the bands of interest from the agarose matrix.Learn how to quickly perform nucleic acid purification using the E-Gel Power Snap Electrophoresis System and an E-Gel CloneWell II gel. Simply load your samples, run the gel, and collect the purified DNA from the recovery wells with a pipet.Find out more at thermofisher.com/clonewell Contents 15'