b'DNA purification in the cloning workflowPurification of PCR productsPCR products taken from agarose gels, PCR tubes, or PCR plates are commonly purified before cloning with restrictionWhat is the intended use of the isolated plasmid?enzymes. We offer several silica-based and magnetic beadbased kits for fast and efficient purification to help youMolecular applications like cloning, get to your next step quickly. PCR, sequencingYes NoThe Thermo Scientific GeneJET Gel Extraction Kit and Invitrogen PureLink Quick Gel Extraction Kit provideGeneJET kitshigher yields and are easier to use than column-based Endotoxin level: 10.0 EU/gkits from other vendors. PCR cleanup with the InvitrogenPrep size: mini, midi, maxiChargeSwitch PCR Cleanup Kit is faster and moreProtocol duration: 1560 min Transfection of robust cell lines like HEK, CHOYield: 0.021 mgcost-effective than cleanup with other magnetic beadbasedCulture volume: 1200 mLkits on the market.Yes NoAdvanced plasmid DNA purification If you are planning to use plasmids to transfect mammalian Transfection of sensitive cells like primary cell cultures, cells, we recommend using a low-endotoxin purification kit likeGibco Expi expression systems, and in vivo usethe Invitrogen PureLink Fast Low-Endotoxin Midi PlasmidPureLink HiPure kitsPurification Kit or an endotoxin-free Invitrogen PureLink ExpiEndotoxin level: 1 EU/g Yes NoPrep size: mini, midi, maxi, mega, gigaplasmid purification kit. With PureLink Expi endotoxin-freeProtocol duration: 90120 minplasmid purification kits, you can obtain high-quality plasmidYield: 0.01514 mg PureLink Expi Endotoxin-Free kits Contact preparations in excellent yields for advanced applicationsCulture volume: 1 mL5 L Endotoxin level: 0.1 EU/g technical support Prep size: maxi, mega, giga thermosher.com/like primary and stem cell transfection, microinjection, geneProtocol duration: 90120 min contacttherapy, and in vivo vaccine research. PureLink Fast Low-Endotoxin kits Yield: 0.415 mgEndotoxin level: 1 EU/g Culture volume: 100 mL5 LPrep size: midi, maxiProtocol duration: 1530 minYield: 0.41.5 mgCulture volume: 30150 mLContents 17'