b'Cloning with synthetic GeneArt Strings DNA Fragments If you lack starting material or dont have enough time to prepare and clone a DNADesigninsert, we can help. With GeneArt Strings DNA Fragments, you can obtain genes that are analogous to error-free PCR products, simply by providing the desired sequence.Using GeneArt Strings DNA Fragments is a time-saving alternative to PCR cloning. They are available in lengths up to 3 kb and are compatible with any downstream cloning method. GeneArt Strings DNA Fragments are particularly useful for assembling large orProduction of DNAOligonucleotide complex genes through Gibson Assembly or type IIs cloning.fragments usingsynthesisYou can easily change a sequence or add restriction sites by typing or clicking a button.GeneArt technologyYou can also increase your level of protein expression using free Invitrogen GeneArt GeneOptimizer software.Gene assemblyOrdering is easy with our online GeneArt dashboard:thermofisher.com/geneartdashboard DirectDirect cloning assemblySequences synthesized with GeneArt Strings DNA Fragments are ready for direct cloning or gene assembly.Find out more at thermofisher.com/strings Contents 7'