b'Selecting the right DNA and RNA isolation kitspreparation SampleFor the quality and performance you need, a full suite of products for DNA and RNA isolation is available for a wide range of sample types, throughputs, and input quantities. To use our online kit selection guide, go to thermofisher.com/rnaselection.Applied Biosystems and Invitrogen technologies for DNA and total RNA isolationProcess a large amountFast isolation of RNA High-throughput purification Process cellsCapabilities oftissue from a variety of samples of RNA andDNA for gene expressionKits TRIzol reagents PureLink kits MagMAX kits Cells-to-CkitsTPrep time 3060 min 20 min 45 min 10 minSample types Most samples, particularly thoseBacteria, liquid, blood, cells, yeast,Blood, plants, saliva, urine, stool,Cultured cells more difficult to lyse plants, tissue soil, plasma, serum*10 8cells, 200 mg of tissue, Starting material 100 mg of tissue or 10 7cells250 mg of plant tissue, 0.2 mL of blood,Variable depending on sample 1100,000 cells5 x 10 8yeast, 10 9bacteria 10 6epithelial cells: 815 gYield 100 mg tobacco leaf: 73 gUp to 350 gVariable depending on sample NA(Variable depending on sample)High-throughput compatible Yes Yes YesTechnology Organic extraction Silica membrane spin column/filter plate Magnetic beads Crude lysate* Specialty kits with optimized chemistry are available for extraction of cell-free DNA/RNA, total RNA, gDNA, and total nucleic acid.Helpful tipIf you are not ready to process your RNA sample, simply store it in Invitrogen RNAlater Stabilization Solution for use at a later time. Visit thermofisher.com/stabilizerna.Learn more at thermofisher.com/rnapreps Contents 11'