b'Transformation (cont.)Medium- and high-throughput transformationPerforming bacterial transformations one by one can be very time-consuming and create a bottleneck in your experimental workflow. There are times when medium- and high-throughput transformation options are desired. Invitrogen MultiShot chemically competent cells provide three flexible product formats to meet your throughput needs. Find out more at thermofisher.com/multishotStripWell format Medium-throughput option Twelve 8-tube strips Suitable for 196 transformations Five E. coli strains availableFlexPlate format High-throughput option 96-well plate separates into 12 x 8-well segmentsDid you know? Manual and automated platform transformations Invitrogen competent cells can be providedSix E. coli strains available in custom configurations per your request. Large and custom volumes as well as multiple formats are at your fingertips. Simply email Cloning96-well plate us at customorders@thermofisher.com. Highest-throughput option Five 96-well plates Available with the TOP10 strain Stable replication of high copy number plasmids46 Contents'