b'Select the Applied Biosystemsthermal cycler thats right for youCloud-enabled Cloud-enabledProFlexPCR System VeritiProThermal Cycler PCR Do you share the device with colleagues? Do you perform a lot of optimizations?Do you expect your throughput needs to change?Do you want to access your instrument remotely?Do you want to access your instrument remotely?Key benefits Ultimate flexibility and throughput Ultimate performanceMax sample throughput 480,000 reactions 384 reactionsMax block ramp rate 6.0C/sec 6.0C/sec6-zone VeriFlex Block on 96-well system6-zone VeriFlex Block on 96-well systemTemperature optimization 2-zone VeriFlex Block on 3 x 32-well systemCompatible with FleetYes YesControl SoftwareDo you require an FDA Class 1/CE-IVD labeled thermal cycler?Visit thermofisher.com/veritidx22 Contents'