b'Type IIS restriction enzymesA specific group of restriction enzymes called Type IIS endonucleases cleave DNA outside of their recognition sequences. In combination with DNA ligase, Type IIS restriction enzymes are utilized to drive the insertion of one or several DNA fragments into a recipient vector without the inclusion of residual restriction enzyme sites and other unwanted DNA sequences at fragment junctions (scarless cloning).Find FastDigest Type IIS enzymes at thermofisher.com/fastdigesttypeiisFor GeneArt Type IIS Assembly Kits, go to thermofisher.com/typeiisCloningContents 41'