b'Sample preparation Automation platformOptimize and automate your DNA and RNA purification workflow with Thermo Scientific KingFisher purification systems. When used with compatible bead-based reagents, such as MagMAX and Dynabeads products, these versatile instruments enable the automation of DNA, RNA, protein, and cell isolation procedures. Learn more and request a demo at thermofisher.com/kingfisher.Thermo Scientific KingFisher Apex Purification SystemThe most modern automated sample purification system includes a touchscreen monitor that lets you edit, create, and run protocols directly on the instrument. Built-in UV decontamination, heating and cooling options, and dual magnet heads allow you to elute directly into storage tubes. This instrument is easyResourcesto use, saves time, and helps ensure consistent results even as laboratory needschange. Navigate through the DNA and RNA support categories below to obtain relevant technical information, view tips and tricks when starting an Thermo Scientific KingFisher Flex Purification System experiment, and find answers to everyday problems.A versatile benchtop automated purification system that processes 24 or 96 samples per run using magnetic bead technology. Gently and efficientlythermofisher.com/napsupportisolate and purify nucleic acids, proteins, and cells from a range of samplethermofisher.com/technicalresourcestypes for a wide variety of downstream applications.thermofisher.com/prepforsuccessthermofisher.com/rnabasicsthermofisher.com/rnahandlingtipsThermo Scientific KingFisher Presto Purification SystemAutomated nucleic acid purification of 24 to 96 samples per run using magneticthermofisher.com/magmaxbead technology. This instrument is specifically designed to be paired with athermofisher.com/dynabeadsliquid handler for high-throughput, fully automated nucleic acid purification. Thermo Scientific KingFisher Duo Prime Purification SystemA versatile compact benchtop sample preparation instrument for the lab that processes 6 or 12 samples per run. It offers consistent extraction and purification of DNA, RNA, proteins, and cells, and includes an ultraviolet lamp for decontamination. The KingFisher Duo Prime system is an affordable choice for academic or small labs.Learn more at thermofisher.com/kingfisher 12 Contents'