b'Applied Biosystems PCR and qPCR plastics are validated and tested for reliability and optimal performance. They are Engineer Approved for use with all Applied Biosystems thermal cyclers and real-time PCR instruments, and are available in a variety of 32-, 48-, 96-, and 384-well plates; tube strips; single tubes; caps; and seals. The table below provides a detailed comparison of each product. Easily find the PCR and qPCR plastics compatible with your instrument using the online selection tool at thermofisher.com/findplastics.Small-scale experiments Complete-workflow experimentsidealDiagnostic procedures and with a few samples Daily experiments for automation automation compatibleSingle tubes, strips, caps, adhesiveMicroAmp EnduraPlate EnduraPlate opticalfilm, and accessories MicroAmp optical microplates optical microplates GPLE* reaction platesFormats Single tubes 32-well96-well 96-wellSingle tubes with caps 48-well Fast 96-well Fast 96-well Fast8-strip tubes with caps 96-well 384-well 384-well PCR12-strip caps 96-well Fast 96-well full skirted384-wellDNA-, RNase-, Yes Yes Yes YesPCR inhibitorfreeColors available Clear, or mixed packs containing red,Clear Single-color packs (red, blue, green,Clearorange, blue, and green yellow, or clear) and 5-plate sampler(one of each color)Barcode available No Yes (1 or 2 sides) Yes (3 sides) Yes (3 sides)Automation compatible Yes Yes* Each lot of reaction plates is certified in an ISO 13485registered facility to be free of DNA, RNase, and PCR inhibitors. Ideal for use in diagnostic procedures.Did you know?Low-profile plastics, also referred to as Fast tubes or plates, are generally required for fast (0.1 mL) thermal blocks. Fast plastics utilize lower volumes (0.1 mL) than the standard (0.2 mL) tubes or plates. The low profile minimizes the air space above the reaction, helping reduce the effects of evaporation and enhancing thermal conductivity. Learn more about PCR and qPCR plastics at thermofisher.com/pcrplastics-education.Contents 25'