b'PCR and qPCR plastics, seals, and accessories Since PCR is a sensitive detection method, PCR plasticsConstruction materials must be of high quality and free of contaminantsApplied Biosystems MicroAmp optical microplates and inhibitors, to help enable optimal performance.are made of polypropylene for optimal transfer of Regardless of the plastics format you select, properthermal energy for efficient PCR. A select medical-grade fit and uniform heat transfer during thermal cyclingpolypropylene is chosen for its exceptional biocompatibility are essential.and inert properties.Manufacturing quality control Applied Biosystems EnduraPlate microplates Applied Biosystems PCR and qPCR plasticare constructed with a stronger polycarbonate frame consumables are manufactured in world-class facilitiesto resist distortion caused by robotic grippers and to dedicated to the production of high-quality molecularbetter tolerate rapid heating and cooling, while retaining thin-walled polypropylene wells for efficient heat transferMicroAmp optical microplatesbiologygrade plastics. After manufacturing, all plastics undergo stringent quality control.to the reaction mixture. The polycarbonate frames PCR of the plates are available in multiple colors to help Integrity testing: Every well of every plate is visuallywith organization and visual monitoring of assays in a inspected and leak tested. This thorough screeninghigh-throughput setting.verifies every well is intact to protect all reactions.Evaporation testing: Samples are run through PCR to test sealing performance. Well liquid volumes are analyzed post-PCR to verify seal integrity. This helps ensure that every production lot conforms to strict tolerances.Biological testing: Our plastics are biologically tested to certify them as free of DNA, RNase, and PCR inhibitors.Polycarbonate (hard shell) frameWe offer General Purpose Laboratory Equipment (GPLE) plastics that are provided with a PCR certificate for yourSkirtconvenience and documentation. Polypropylene wellsEnduraPlate microplate forhigh-throughput robotic applicationsFind out more at thermofisher.com/pcrplastics 24 Contents'