b'Reverse transcription is the synthesis, by reverse transcriptase, of complementary DNA (cDNA) using single-stranded RNA as a template. The cDNA can be used as a template for PCR amplification, cDNA library construction, RNA sequencing, and more. Selecting the right reverse transcriptase is critical to detecting low-abundance RNAs in a sample and obtaining high yields of full-length cDNA.Find technical resources on reverse transcription atthermofisher.com/rteducation14'