b'PCR and qPCR plastics, seals, and accessories Since PCR is a sensitive detection method,Construction materials PCR plastics must be of high quality and free ofApplied Biosystems MicroAmp optical contaminants and inhibitors, to help enable optimalmicroplates are made of polypropylene for optimal performance. Regardless of the plastics format youtransfer of thermal energy for efficient PCR. A select select, proper fit and uniform heat transfer duringmedical-grade polypropylene is chosen for its thermal cycling are essential.exceptional biocompatibility and inert properties.Manufacturing quality control Applied Biosystems EnduraPlate microplates Applied Biosystems PCR and qPCR plasticare constructed with a stronger polycarbonate consumables are manufactured in world-classframe to resist distortion caused by robotic grippers facilities dedicated to the production ofand to better tolerate rapid heating and cooling,MicroAmp optical microplates.high-quality molecular biologygrade plastics.while retaining thin-walled polypropylene wells for PCR After manufacturing, all plastics undergo stringentefficient heat transfer to the reaction mixture. The quality control.polycarbonate frames of the plates are available in multiple colors to help with organization and visual Integrity testing: Every well of every plate is visuallymonitoring of assays in a high-throughput setting.inspected and leak tested. This thorough screening verifies every well is intact to protect all reactions.Evaporation testing: Samples are run through PCR to test sealing performance. Well liquid volumes are analyzed post-PCR to verify seal integrity. This helps ensure that every production lot conforms to strict tolerances. Polycarbonate (hard shell) frameBiological testing: Our plastics are biologicallySkirttested to certify them as free of DNA, RNase,Polypropylene wellsand PCR inhibitors. We offer General Purpose Laboratory Equipment (GPLE) plastics that are provided with a PCR certificate for your convenience and documentation.EnduraPlate microplate forhigh-throughput robotic applications.Find out more at thermofisher.com/pcrplastics 24'