b'Electrophoresis reagents For pouring your own agarose gels, choosingUltraPure reagents for electrophoresishigh-quality agarose, optimized DNA ladders, andInvitrogen UltraPure reagents are specifically improved DNA stains can help you achieve optimalformulated to meet your nucleic acid analysis and electrophoresis results.purification needs. UltraPure agarose and reagents are made from highly pure biochemicals for DNA stains maximum reliability and superior performance. Detection of nucleic acid samples in gels can be improved using fluorescent dyes that are saferFind out more at thermofisher.com/ultrapureand/or more sensitive than ethidium bromide. The Invitrogen SYBR Safe, SYBR Green I, andDNA laddersSYBR Gold stains provide greater safety and/ Invitrogen DNA ladders are available in a wide or sensitivity with lower background fluorescencevariety of size ranges (10 bp to 15 kb) and formats than the conventional ethidium bromide stain.for different applications. To create DNA ladders of superior quality, each fragment is purified Find out more at thermofisher.com/stains individually using proprietary chromatography-based technology. Our DNA ladders are stable during prolonged storage at room temperature and after multiple freeze-thaw cycles. ElectrophoresisFind out more at thermofisher.com/laddersFluorescent nucleic acid gel stainsStandardSaferEnhancedUltimate detection detection detection detection Did you know?UltraPure ethidiumSYBR Safe SYBR Green IChromatographically purified nucleic acid bromide stain stain SYBR Gold stain fragments are considered the gold standard Sensitivity (dsDNA) SensitiveSensitiveHighly sensitive Ultrasensitive for ladders, since the technology provides (1 ng) (1 ng) (60 pg) (25 pg) higher control over quality, banding pattern, Less hazardous and more intensity, and quantity for ladder composition.environmentally friendlyImproved cloningLearn more at thermofisher.com/ efficiencyna-electrophoresis-education36'