b'PCR primers and DNA oligos Good design (i.e., good sequence selection)Custom DNA oligosand high-quality primers are critical to yourInvitrogen custom DNA oligos are synthesized PCR reactions. In general, a length of 1830on a highly automated, computer-controlled nucleotides for primers is optimal. The meltingsystem, followed by rigorous quality control. Mass temperatures (T ) of the primers should bespectrometry and capillary electrophoresis areHelpful tipmbetween 65C and 75C, and within 5C ofperformed for short and long oligos, respectively,If the T mof your primer is very low, try to each other. to help ensure the quality of the process andfind a sequence with higher GC content; end products.alternatively, the length of the primer can be OligoPerfect primer designer extended. For more tips on primer design,Whether your are performing PCR, cloning, orThe appropriate synthesis scale and purificationgo to thermofisher.com/primerdesigncapillary electrophoresis (CE) sequencing, takefor your application depend on the nature of your advantage of the benefits offered by our robustdownstream applications. Choose the right oligos PCR and easy-to-use Primer3-based Invitrogenand purification methods for your applications. OligoPerfect Designer. Speed updesign primers for up to Processing option Desalted HPLC PAGE50 genes at the same time Oligos25 nmol10 mol50 nmol10 mol50 nmol10 molStore your dataability to save5100 bp755 bp7100 bpyour projects85% full-length sequence90% full-length sequenceStandard PCR Work smarterrecognizes .txt andSpecialty PCR.fasta file typesCloningOrder with easeseamlessly integratesIn addition to standard delivery, next-day delivery is also available in most regions where local oligo with the Invitrogen ordering portal manufacturing exists. For more on ordering information, yield guarantees, designing tools, technical resources, protocols, and FAQs, go to thermofisher.com/oligosTry the OligoPerfect Designer at thermofisher.com/oligoperfect-designerFind out more at thermofisher.com/oligos 30'