b'Cloud-enabledCloud-enabled MiniAmpPlus andSimpliAmpThermal Cycler MiniAmpThermal Cyclers Automated Thermal Cycler Do you need an intuitive interface? Do you want to place yourPCRDo you want an instrument with just the features Do you train new technicians often? needed for routine PCR? instrument on a robotic platform Do you want to access your instrument remotely? Do you want to access your instrument remotely? now or in the future? Elegantly simple and preciseRoutine PCR, elevatedDesigned for easy robotic integration 96 reactions96 reactions384 reactions 4.0C/secMiniAmp Plus: 3.5C/sec; MiniAmp: 3.0C/sec3.5C/sec3-zone VeriFlex Block on 96-well system MiniAmp Plus: 3-zone VeriFlex Block Noneon 96-well system MiniAmp: noneYes Yes YesFind out more at thermofisher.com/thermalcyclers 23'