b'I nnovations for superior PCRPCR enzymes and reagents are continually being improved to help you get to your research destination faster. For example, the latest Platinum DNA polymerases are designed with the following key innovative features.More robust and versatileAdvanced enzymatic engineering and methodology provide DNA polymerase with fast cycling, high tolerance of PCR inhibitors, and efficient amplification of challenging DNA like GC-rich sequences. These features help you amplify DNA targets confidently with speed and simplicity.Find out more at thermofisher.com/platinumenzymesPCRUniversal primer annealingThe innovative Platinum PCR buffers enable universal primer annealing at 60C. This design allows you to co-cycle different PCR assays (instead of running them sequentially), drastically reducing tedious optimization steps and saving time.Find out more at thermofisher.com/universalannealingDirect gel loading The latest Platinum DNA polymerases are available in a green buffer format that allows direct gel loading and eliminates tedious steps of dye addition, helping reduce pipetting errors. DNA migration is easily tracked with two dyes (blue and yellow) that are readily visible during electrophoresis (the lanes for 5 and 15 min in the figure to the right).29'