b'Selecting the right DNA isolation kitspreparation SampleTo suit your specific DNA isolation needs, a comprehensive portfolio is available offering three highly developed purification technologies: silica membrane, anion-exchange resin, and switchable surface charge. Refer to the following tables to determine which purification kit is appropriate for your DNA type and sample type. To use our online kit selection guide, go to thermofisher.com/dnaselectionApplied Biosystems and Invitrogen technologies for gDNA isolationLow throughput Medium throughput and High throughput andHigh throughput andThroughput/format or manual (organic) spin-column technology 96-well filter plate magnetic bead technologyKits DNAzol reagents PureLink kits PureLink Pro gDNA kits MagMAX DNA kits and Dynabeads productsTissue DNAzol Reagent PureLink Genomic PureLink Pro 96 Genomic MagMAX DNA Multi-Sample KitDNA Mini Kit DNA Purification KitCells DNAzol Reagent PureLink Genomic PureLink Pro 96 Genomic MagMAX DNA Multi-Sample KitDNA Mini Kit DNA Purification KitBlood DNAzol BD Reagent PureLink Genomic PureLink Pro 96 Genomic MagMAX DNA Multi-Sample Ultra DNA Mini Kit DNA Purification Kit 2.0 KitPlant Plant DNAzol Reagent PureLink Genomic Plant PureLink Pro 96 Genomic MagMAX Plant DNA Isolation KitDNA Purification Kit DNA Purification KitBuccal swabs Not recommended PureLink Genomic PureLink Pro 96 Genomic MagMAX DNA Multi-Sample Ultra DNA Mini Kit DNA Purification Kit 2.0 KitPureLink Microbiome PureLink Pro 96 Genomic MagMAX DNA Multi-Sample Bacteria DNAzol Reagent DNA Purification Kit DNA Purification Kit Ultra 2.0 Kit, Dynabeads MyOne Streptavidin productsPureLink Viral RNA/DNA PureLink Pro 96 Viral MagMAX Viral Nucleic Acid Kit, Virus Not recommended Mini Kit RNA/DNA Purification Kit Dynabeads MyOne Streptavidin productsCompatibilityScalable and automatableThermo Scientific KingFisher instrument qPCRNGSFind out more at thermofisher.com/gdnaprep 9'