b'E-Gel Power Snap Electrophoresis SystemTo help reduce user errors and workflow time, the Invitrogen E-Gel Power Snap Electrophoresis System integrates rapid, real-time nucleic acid analysis with high-resolution image capture. The system offers: Faster analysisSimple operation Safer workflow go from sample loadingintuitive user interface withminimize handling of to image capture in aslarge touchscreen andhazardous chemicalslittle as 18 minutes integrated operating systemFind out more at thermofisher.com/powersnapElectrophoresisE-Gel system for high-throughput electrophoresisHigh-throughput electrophoresis is often performed for high-volume analysis of PCR products, plasmid preparations, and restriction digests. Gel runs and analysis can be accelerated using the E-Gel 48 and E-Gel 96 precast gels and expandable Invitrogen E-Base Electrophoresis System. The integrated design of the E-Gel Mother E-Base and Daughter E-Base devices saves space and allows up to 384 samples to run at one time.Find out more at thermofisher.com/egel-highthroughput35'