b'PCR reagents DNA polymerase is an essential component forFidelityPCR because of its key role in synthesizingThe fidelity, or proofreading capability, of a DNA new DNA strands. Because of the sensitive andpolymerase is based on its 3 to 5 exonuclease specific nature of PCR, it is important to chooseactivity, which corrects misincorporated high-quality enzymes and reagents to producenucleotides. This function is critical in applicationsDid you know?optimal results. The following are things tosuch as cloning, sequencing, and site-directedThe residual bacterial DNA in recombinant consider when choosing PCR enzymes. mutagenesis, for accurate replication ofPCR enzymes poses challenges in microbial DNA sequences.genome analysis, such as accurately Specificity detecting bacterial strains by 16S rRNA Nonspecific amplification is one of the majorProcessivity gene sequences. To enable confidence and hurdles in PCR, since it can drastically impactA DNA polymerases processivity is defined assuccess in microbial PCR assays, choose PCR enzymes with controlled low levels of yield and sensitivity of target amplification. Onethe number of nucleotides being incorporated inresidual bacterial and human genomic DNA. PCR way to help reduce nonspecific amplification isa single binding event. This property often reflects through the use of a hot-start DNA polymerase,synthesis rate and speed, as well as affinityFind out more at which utilizes an antibody or chemical modificationfor its substrates. Therefore, highly processivethermofisher.com/broad-range-pcrso that the polymerase becomes active onlyDNA polymerases are beneficial to amplify at the high temperature of the denaturationchallenging templates such as long, GC-rich, or step. In addition to improving specificity, a hot- inhibitor-containing DNA.start DNA polymerase increases yield and allows convenient room temperature setup forPrimer annealing temperaturehigh-throughput applications. The primer annealing temperature of each DNAHelpful tipfragment to be amplified often needs optimizationDirect PCR is a way to help simplify PCR Thermostability when designing a PCR protocol. To help simplifyexperiments, save time, and prevent sample Since thermal cycling is a key feature of theannealing and enable co-cycling of PCR assays,loss in the workflow. Direct PCR allows you conditions that enable the repetitive chain reactionconsider a DNA polymerase with a reaction bufferto amplify target sequences directly from the of amplifying DNA, thermostability of the DNAthat allows a universal annealing temperature ofsamples without the need to first isolate and polymerase to be used is also an important60C for primers. purify the DNA.feature. Highly thermostable DNA polymerasesFind out more atare recommended for amplifying GC-rich orthermofisher.com/direct-pcrlong templates that often require prolonged high-temperature reactions.Find out more at thermofisher.com/pcrenzymes 26'