b'Simplify electrophoresis with E-Gel precast agarose cassettes E-Gel precast gelsUsing precast agarose gels can simplify the nucleic acid electrophoresis workflow. E-Gel precast gels are self-contained and ready for use with the agarose, electrodes, and the DNA stain packaged inside a disposable cassette. There are no gels to pour, buffers to make, staining or destaining steps to perform, or gel boxes to assemble. Just load your samples and run. E-Gel precast gels offer excellent resolution and clarity in 18 minutes and are ideal for analyzing PCR products, restriction digests, plasmid preparations, and genotyping products. To help simplify cloning workflows, Invitrogen E-Gel CloneWell II gels use a double-comb design to enable recovery of purified DNA for downstream applications, without the need for additional purification kits or steps.Find out more at thermofisher.com/egelElectrophoresisE-Gel DNA ladders Accurate analysis of electrophoresis bands often depends on the DNA ladder you choose for your gel run. For optimal performance on E-Gel precast agarose gels, Invitrogen E-Gel DNA ladders are formulated with chromatography-purified DNA fragments in ready-to-use loading buffer. The chromatography-based purificationHelpful tipmethod results in exceptional purity and quality of the DNA fragments, while anE-Gel precast gels are available in a variety of formats for optimal buffer formulation reduces dye masking and helps improve ladder migrationroutine and high-throughput applications, with different stains for more accurate analysis.(see page 36) and agarose percentages (0.8%, 1.2%, 2%, and 4%). To find the right gel for your needs, see the selection guide at thermofisher.com/egelselectionFind out more at thermofisher.com/egel-laddersFind out more at thermofisher.com/egels34'