b'Select the Applied Biosystems thermal cycler thats right for you Cloud-enabledProFlexPCR System Veriti Thermal CyclerPCR Do you share the device with colleagues? Do you perform a lot of optimizations?Do you expect your throughput needs to change? Do you require an FDA Class 1/CE-IVD Do you want to access your instrument remotely? labeled device?Key benefitsUltimate flexibility and throughputProven reliability, precise PCR optimization Max sample throughput480,000 reactions384 reactions Max block ramp rate6.0C/sec5.0C/secTemperature optimization 6-zone VeriFlex Block on 96-well system6-zone VeriFlex Block on 96-well system2-zone VeriFlex Block on 3 x 32-well systemCompatible with FleetYesControl SoftwareDo you require an FDA Class 1/CE-IVD labeled thermal cycler?Visit thermofisher.com/veritidx22'