b'SeparateTransferDetectFast protein transferiBlot2 Gel Transfer Device The Invitrogen iBlot 2 Gel Transfer DeviceiBlot 2 Starter Kit (Cat. No. IB21001S)is our premium western blot transfer device, delivering high performance and convenience. A unique, innovative system, the iBlot 2 device Minimal preparation and cleanuptransfer utilizes optimized, preassembled transfer stacksstacks streamline transfer setup and teardownwith transfer buff er incorporated into gel matrices, so theres no need to prepare messy transfer Convenienttouchscreen interface, buff ers. Just insert your gel and go. preprogrammed and optimized transfer protocols, and prepackaged ready-to-useEngineered for rapid transferthe shortstacks for transferring midi and mini blot formats distance between electrodes, along with high(1 midi blot or up to 2 mini blots at a time) fi eld strength and current, reduces transfer time to just 7 min Find out more at thermofi sher.com/iblot2A B CBolt Bis-Tris Plus, NCNuPAGE Bis-Tris, NC Novex Tris-glycine, NCD E FBolt Bis-Tris Plus, PVDF NuPAGE Bis-Tris, PVDF Novex Tris-glycine, PVDFMembranes processed on the iBlot 2 Gel Transfer Device show consistent transfer across various protein gel chemistries to both nitrocellulose (NC) and PVDF membranes. Total cell extracts from A431 cells were transferred to NC (AC) or PVDF (DF) membranes from Invitrogen Bolt 412% Bis-Tris gels, Invitrogen NuPAGE 420% Bis-Tris gels, and Invitrogen Novex 420% Tris-glycinegels. 10'