b'SeparateTransferDetectStunningly easy western blot imagingiBright Imaging Systems Simpli ed western blot and gel imagingiBright Imaging Systems provide:Experience an easier time capturing and analyzing Push-button, optimized exposuredata from gels and western blots with InvitrogenSmartExposure acquisition technology for the iBright Imaging Systems. Designed with arapid determination of optimal exposure times streamlined, intuitive interface and workfl ows,helps minimize the need to repeat exposures to iBright Imaging Systems are easy to use forget the desired signalresearchers of all experience levels.Powerful 9.1 megapixel (MP) cameraThere are three models in the iBright Imagingcapture crystal-clear images with robust Systems family: The iBright CL750 Imagingimaging potentialSystem, the iBright CL1500 Imaging System, Advanced automated featuresautomatic and the iBright FL1500 Imaging System. Thesample rotation, auto-zoom, auto-focus, and iBright CL750 Imaging System off ers the coreautomatic onboard data analysis provide a essential western blot and gel imaging functionssmooth imaging experienceand makes the transition from the darkroom and fi lm easy. The iBright CL1500 Imaging System Five-channel fl uorescent blottingmultiplex expands application support and has many of thewith the fi ve fl uorescence channels of the high-performance specifi cations of our premieriBright FL1500 model; capture up to four iBright FL1500 model. The iBright FL1500 Imagingproteins in a single blot for more meaningful and System features maximum application support,representative experimentsincluding fl uorescent western blot imaging with up Compliance supportall models off ered with to four fl uorescence channels at a time.21 CFR Part 11 compliance support software packages to set up and control security, audit, and e-signature settingsCapture your data faster and more easily than ever before. Find out more at thermofi sher.com/ibright14'