b'SeparateTransferDetectAutomated western blot processingBandmate Automated Western Blot ProcessorThe Invitrogen Bandmate Automated Western Blot Processor is a programmable blot rocking system that automates the tedious hands-on blocking, washing, and antibody incubation steps of western blot processing. Minimal eff ort is required to set up the Bandmate device to process up to 2 midi blots or 4 mini blots using your current optimized reagents and protocols for blot processing, freeing up time for other important tasks.The Bandmate Automated Western Blot Processor off ers: Load-and-go setupprepare block, wash, and antibody solutions, load into the machine, select a program, and walk away Works with traditional western blotting protocolsno need to adapt from current protocols and no specialized reagents required; program the timing of steps based on preference or utilize preprogrammed options Antibody recoverycollection tubes can recover antibody for reuse in future experiments if desiredExplore more at thermofi sher.com/bandmateComparison of mini blots processed with the Bandmate Automated Western Blot Processor vs. manual processing (probing and washing steps performed in a tray and on a shaker platform). The blots processed with the Bandmate device show comparable intensity levels across multiple probed targets compared to the blots processed with a manual processing procedure.23'